Learn More
Corning™ Plain Reusable Short Stem Plastic Funnels
Durable polypropylene funnels are translucent and allow safe and easy transfers of solids or liquids from one vessel to another.
Brand:  Corning™ 6120P-150
 View more versions of this product
Product Code. 16546834
  • 411.00 DKK / Pack of 12
Estimated Shipment: 11-09-2025
to see stock.
Explore more special offers
Description
Allows safe and easy transfer of solids or liquids from one vessel to another.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15 mm
220 mm
95 mm
Polypropylene
Constructed of durable translucent Polypropylene
Product Title
Corning™ Plain Reusable Short Stem Plastic Funnels > 150 mm