Learn More
Corning™ 384-Well Clear Polystyrene Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
Brand: Corning™ 3680
4725.80 DKK valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Specifications
Standard | |
0.06 cm2 | |
Clear | |
384 | |
Sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
With Lid | |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. | |
20/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
0Hours
6Minutes
55Seconds
1