Learn More
Corning™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand:  Corning™ 3843
Product Code. 11389514
  • 15132.83 DKK / Pack of 100
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Description
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Standard
    0.32 cm2
    White, Clear Bottom
    96
    Sterile
    Flat Bottom (F-well)
    100/Cs.
    Clear
    With Lid
    8 x 12
    Polystyrene
    75 to 200 μL
    Poly-D-Lysine
    Easy transition into existing automation and readers, reduced evaporation of well contents, enhanced cell attachment, enhance luminescent signals and have low background luminescence and fluorescence
    20/Pk.
    Documents
    Product Information
    Product Title
    Corning™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate > White and Clear; Standard, with Lid; Yes; Poly-D-Lysine; Flat