Learn More
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes
With white rubber liner
Brand: Corning™ 99999-15
708.78 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Made of a phenolic compound. These caps have a white rubber liner.
Specifications
15-415 | |
Screw Cap | |
Phenolic | |
16 mm | |
1000/Cs. | |
Culture Tubes |
Rubber | |
Autoclavable | |
Black | |
1000/Pk. | |
Rubber |
Product Content Correction
Product Title
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes > 15-415
Spot an opportunity for improvement?Share a Content Correction