Learn More
Corning™ Counting Chamber
Corning counting chamber is a reusable glass cell counting slide for the Corning cell counter.
Brand:  Corning™ 480200
962.58 DKK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 15968746
  • 962.99 DKK / Each
Estimated Shipment: 10-09-2025
to see stock.
Explore more special offers
Description
This gridless slide has a 0.1 mm depth same as a Neubauer Hemocytometer and comes standard with two high quality cover slips. These slides have a notch for easy loading of cell suspension.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gridless
0.1 mm
Corning™ Cell Counter
Product Title
Corning™ Counting Chamber > Gridless; 0.1 mm depth; For counting chamber