Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
46.95 DKK - 48.85 DKK
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | Polyethylene |
Color | White |
Ankle Style | Elastic |
Neck Style | Hooded |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15310541
![]() |
DuPont™
D14886039 |
Small |
47.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
15320541
![]() |
DuPont™
D14886047 |
Medium |
48.83 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
15330541
![]() |
DuPont™
D14886050 |
Large |
48.83 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
15360541
![]() |
DuPont™
D14886081 |
3X-Large |
48.85 DKK
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
15350541
![]() |
DuPont™
D14886075 |
2X-Large |
46.95 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
15340541
![]() |
DuPont™
D14886064 |
X-Large |
46.95 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
Description
- Robust yet lightweight
- Elasticated face, wrists, waist and ankles
- Antistatic treatment on the outside
- Stitched external seams in yellow for visual identification and differentiation
Specifications
Hooded Coveralls | |
White | |
Hooded | |
Zipper | |
Category III, Type 5 and 6 |
Polyethylene | |
Elastic | |
2-panel | |
Stitched | |
(EU) 2016/425, EN 1073-2, EN 1149-1 |