Learn More
Novus Biologicals™ CpG oligodeoxynucleotides Mouse
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
Specifications
Specifications
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Quantity | 0.025 mg |
For Use With (Application) | Activation of mouse TLR9 |
Type | CpG Oligodeoxynucleotides |
Product Content Correction
Product Title
Novus Biologicals™ CpG oligodeoxynucleotides Mouse >
Spot an opportunity for improvement?Share a Content Correction