Learn More
Novus Biologicals™ CpG oligodeoxynucleotides Mouse
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
Concentration | 1.0 mg/mL |
Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
Quantity | 0.1 mg |
For Use With (Application) | Mouse CpG ODNs Are Used for Activation of Mouse TLR9 (at a Range of 5-20 ÎĽg/mL The TLR9/NF-kB/SEAPorter HEK 293 Cell Line is a Useful Positive Control Model System for Studying hTLR9 Activation. We Typically Use a 96-Well Plate Format Assay Using This Reporter Cell Line.) |
Target | CpG Oligodeoxynucleotides Mouse |
Type | CpG Oligodeoxynucleotides |
Product Content Correction
Product Title
Novus Biologicals™ CpG oligodeoxynucleotides Mouse >
Spot an opportunity for improvement?Share a Content Correction