Learn More
JULABO™ CR Tubing
Original high quality JULABO™ accessories for your temperature control unit.
Brand: JULABO™ 8930012
This item is not returnable.
View return policy
Description
- 1 m CR tubing
- -30...+120°C
- 12 mm ID
- max. 1.2 bar
- Chloroprene
- Suitable for JULABO™ baths
Specifications
CR Tubing | |
-30°C to +120°C | |
CR tubing 12 mm I.D. |
CF30, CF31, CF40, CF41 Cryo-compact circulator, F25-HE refrigerated/heating circulator | |
CR Tubing | |
1 m |