missing translation for 'onlineSavingsMsg'
Learn More

CRMP1 Rabbit anti-Human, Rat, Clone: 1U7K3, Novus Biologicals™

Rabbit Monoclonal Antibody

1460.01 DKK - 3692.13 DKK

Specifications

Antigen CRMP1
Clone 1U7K3
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18310597
View Documents Promotion Details
Bio-Techne
NBP3-16120-20UL
20 μg
1460.01 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
18378655
View Documents Promotion Details
Bio-Techne
NBP3-16120-100UL
100 μg
3692.13 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CRMP1 Monoclonal antibody specifically detects CRMP1 in Human, Rat samples. It is validated for Western Blot
Specifications

Specifications

CRMP1
Western Blot
Unconjugated
Rabbit
Neuroscience
PBS, 0.05% BSA, 50% glycerol, pH7.3
1400
IgG
Affinity purified
1U7K3
Monoclonal
Purified
RUO
Human, Rat
collapsin response mediator protein 1DRP1, CRMP-1, dihydropyrimidinase-like 1, dihydropyrimidinase-related protein 1, DPYSL1DRP-1dihydropyrimidinase related protein-1, ULIP3, ULIP-3, Unc-33-like phosphoprotein 3
A synthetic peptide corresponding to a sequence within amino acids 450-550 of human CRMP1 (Q14194).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications