Learn More
Tocris Bioscience™ CRT 0066101
Potent protein kinase D (PKD) inhibitor
Brand:  Tocris Bioscience™ 4975/10
2210.74 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18727286
  • 2800.00 DKK / 10mg
Explore more special offers
Description
Description
CRT 0066101 is a potent inhibitor of protein kinase D (PKD); inhibits all PKD isoforms (IC50 values are 1, 2 and 2.5 nM for PKD1, PKD3 and PKD2 respectively). Exhibits selectivity for PKD against a panel of >90 protein kinases, including PKCα, MEK, ERK, c-Raf and c-Src. Reduces proliferation and cell viability of pancreatic cancer cells expressing moderate levels of endogenous PKD1/2. Orally bioavailable.
Specifications
Specifications
>98%
CRT 0066101
CRT 0066101
10 mg
411.33
Special Offers
For Research Use Only