Learn More
Thermo Scientific™ CryoMed™ Sensors
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Brand: Thermo Scientific™ 4000385
4231.42 DKK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Description
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:- Thermocouple sensors for cryo tubes, sheath straw and vial samples
- Includes male plug
Recommended for:
Controlled-rate freezing applications
Specifications
Thermocouple Sensor | |
7.06 cu. ft. | |
1.2/2.0 Cryo Tubes in CryoMed | |
Cryo Tube, Sheath Straw, Vial |
Thermocouple Sensor | |
2 mL | |
Male |
Promotions
-
Multiple Manufacturers
Illuminating Protein Pathways
Shining a light on the tools that drive protein discovery.Promo Code: 24148Expires: 2025-08-22
New Offer Save Now