missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing without Configuration
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing without Configuration handles virtually all inorganic chemicals found in the lab.
Brand: Saint Gobain Life Sciences 070534-07L-ND
This item is not returnable.
View return policy
Description
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
- Resistant to flex fatigue and abrasion
- Less permeable than rubber tubing
- For use with virtually all inorganic chemicals found in the lab

Specifications
USP Class VI | |
0.64 mm | |
0.09 in. | |
32.8 ft. | |
0.04 in. | |
For sample and reagent dispensing, biological and chemical assays, and water and soil analysis | |
-50°C | |
0.64 x 0.90 mm | |
55 (Shore A) | |
165.2°F | |
Tubing |
0.03 in. | |
Tygon R3607 | |
2.24 mm | |
10 m | |
0.90 mm | |
-58°F | |
1 Ea. | |
Orange/White | |
Flexible | |
74°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction