missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Culti-Loops™ Eikenella corrodens ATCC™ BAA-1152™ Product Code.: 13286159

Thermo Scientific™ Culti-Loops™ Eikenella corrodens ATCC™ BAA-1152™

Product Code. 13286159
5 Loops/Pk., Pack of 5
Click to view available options
Quantity:
5 Loops/Pk.
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 13286159

Brand: Thermo Scientific™ R4601947

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® BAA-1152™*
Certifications/Compliance Culti-Loops are CE Marked.
Description Eikenella corrodens
Detects Eikenella corrodens
Format Culti-Loops
Product Type Culti-Loops Quality Control Organism
Target Organism Class Fastidious
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops/Pk.
Strain Designations Vitek #101889 {NSB #81355]
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number242644 Certificate TypeCertificate of Analysis Date25/03/2025 Product CodeR4601947
Lot Number188125 Certificate TypeCertificate of Analysis Date08/11/2024 Product CodeR4601947
Lot Number149669 Certificate TypeCertificate of Analysis Date09/08/2024 Product CodeR4601947
Lot Number134366 Certificate TypeCertificate of Analysis Date28/06/2024 Product CodeR4601947
Lot Number714636 Certificate TypeCertificate of Analysis Date16/04/2024 Product CodeR4601947

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Culti-Loops™ Eikenella corrodens ATCC™ BAA-1152™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.