Learn More
Thermo Scientific™ Culti-Loops™ Morganella morganii subsp. morganii ATCC™ 25830™
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

Specifications
Specifications
ATCC® | 25830TM* |
Description | Morganella morganii subsp. morganii ATCC™ 25830™ |
Format | Culti-Loops |
Product Type | Culti-Loops Quality Control Organism |
Target Organism Class | Gram-negative Rods |
Quantity | 5 Loops/Pack |
Documents
Scientific Resources
Brochures
Product Information
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.