Learn More
Ambion™ Custom RNA, Stealth, Desalted
Custom RNA, Stealth, Desalted
Brand: Ambion™ 100001855
This item is not returnable.
View return policy
Description
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
Specifications
Room temperature | |
RNA DSL In-vivo, 100nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation | |
RNA DSL In-vivo | |
100 nmol |
Product Content Correction
Product Title
Ambion™ Custom RNA, Stealth, Desalted > 100 nmol
Spot an opportunity for improvement?Share a Content Correction