Learn More
Tocris Bioscience™ CX 614
AMPAkine; positive allosteric modulator of AMPA
Brand: Tocris Bioscience™ 5149/50
5379.56 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
CX 614 is an AMPAkine; positive allosteric modulator of AMPA. Prototypical benzamide which occupies subsite A only. Modulates channel deactivation and desensitization.Specifications
>99% | |
CX 614 | |
CX 614 |
50 mg | |
247.25 |
For Research Use Only