Learn More
Tocris Bioscience™ CX 614
AMPAkine; positive allosteric modulator of AMPA
Brand:  Tocris Bioscience™ 5149/10
1316.60 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18765329
  • 1675.00 DKK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
CX 614 is an AMPAkine; positive allosteric modulator of AMPA. Prototypical benzamide which occupies subsite A only. Modulates channel deactivation and desensitization.
Specifications
Specifications
>99%
CX 614
CX 614
10 mg
247.25
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only