missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals CX3CR1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00001524-B01P
This item is not returnable.
View return policy
Description
CX3CR1 Polyclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
CX3CR1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
NP_001328.1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
Unconjugated | |
PBS (pH 7.4) | |
Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKBRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28C-X3-C CKR-1 | |
CX3CR1 (NP_001328.1, 1 a.a. - 355 a.a.) full-length human protein. MDQFPESVTENFEYDDLAEACYIGDIVVFGTVFLSIFYSVIFAIGLVGNLLVVFALTNSKKPKSVTDIYLLNLALSDLLFVATLPFWTHYLINEKGLHNAMCKFTTAFFFIGFFGSIFFITVISIDRYLAIVLAANSMNNRTVQHGVTISLGVWAAAILVAAPQFMFTKQKENECLGDYPEVLQEIWPVLRNVETNFLGFLLPLLIMSYCYFRIIQTLFSCKNHKKAKAIKLILLVVIVFFLFWTPYNVMIFLETLKLYDFFPSCDMRKDLRLALSVTETVAFSHCCLNPLIYAFAGEKFRRYLYHLYGKCLAVLCGRSVHVDFSSSESQRSRHGSVLSSNFTYHTSDGDALLLL | |
0.05 mg | |
GPCR | |
1524 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |