missing translation for 'onlineSavingsMsg'
Learn More

CXCL8/IL-8 Mouse anti-Bovine, Canine, Clone: MT8H6, Novus Biologicals™

Mouse Monoclonal Antibody

2404.91 DKK - 8120.74 DKK

Specifications

Antigen CXCL8/IL-8
Clone MT8H6
Concentration 0.5 mg/ml
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18337926
View Documents Promotion Details
Novus Biologicals
NBP3-18279-250UG
250 μg
2404.91 DKK
250µg
Please to purchase this item. Need a web account? Register with us today!
18310894
View Documents Promotion Details
Novus Biologicals
NBP3-18279-1000UG
1000 μg
8120.74 DKK
1000µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Bovine, Canine samples. It is validated for ELISA
Specifications

Specifications

CXCL8/IL-8
0.5 mg/ml
Monoclonal
Purified
RUO
Bovine, Canine
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Recombinant human CXCL8/ IL-8
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
MT8H6
ELISA
Unconjugated
Mouse
Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
0.2 um filtered PBS
3576
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications