Learn More
WTW™ Cyanide Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 250492
1253.44 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
DIN 38 405 D13/ISO 6703 | |
Measurement of Cyanide in wastewater from electroplating and metal processing works, drinking water | |
Cuvettes measuring 10mm, 20mm and 50mm | |
0.002 to 0.500 mg/L |
25 mL | |
PhotoLab S12, PhotoLab Spectral | |
100 Pack | |
Test powder |
Product Content Correction
Product Title
WTW™ Cyanide Test Kits > Detection Range: 0.002 to 0.500mg/L
Spot an opportunity for improvement?Share a Content Correction