Learn More
MedChemExpress Cyclo(his-pro)
MedChemExpress Cyclo(his-pro) (Cyclo(histidyl-proline)) is an orally active cyclic dipeptide structurally related to tyreotropin-releasing hormone. Cyclo(his-pro) could inhibit NF-κB nuclear accumulation. Cyclo(his-pro) can cross the brain-blood-barrier and affect diverse inflammatory and stress responses.
Brand:  MedChemExpress HY-101402
Encompass_Preferred
Product Code. 16482726
  • 3021.37 DKK / 25mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
53109-32-3
Neuroscience-Neuromodulation
Natural Products
Room temperature
Cyclo(histidyl-proline)∣ Histidylproline diketopiperazine
Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
234.25 g/mol
25 mg
H2O : ≥ 30 mg/mL (128.07 mM)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.