Learn More
RETSCH Cyclones
20555.00 DKK - 29125.00 DKK
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
11737404
|
RETSCH
22.935.0014 |
1 Sample Bottle 250mL and 1 Sample Bottle 500mL, Filter Bag |
20555.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
11984272
|
RETSCH
22.935.0017 |
1 Sample Bottle 250mL and 1 Sample Bottle 500mL, Filter Bag, Connection for Vacuum Cleaner |
23615.00 DKK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
12652705
|
RETSCH
22.935.0018 |
3 L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner |
29125.00 DKK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
11994272
|
RETSCH
22.935.0019 |
5L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner |
29125.00 DKK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
Specifications
Cyclone | |
For ZM 200 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
RETSCH Cyclones