missing translation for 'onlineSavingsMsg'
Learn More
Cyclooctane, 99+%, Thermo Scientific Chemicals Product Code.: 11328106

Cyclooctane, 99+%, Thermo Scientific Chemicals

Product Code. 11328106
50 mL
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy

Product Code. 11328106

Brand: Thermo Scientific Alfa Aesar L15000.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 292-64-8 | C8H16 | 112.216 g/mol

Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292-64-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00004162
InChI Key WJTCGQSWYFHTAC-UHFFFAOYSA-N
Synonym octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
PubChem CID 9266
IUPAC Name cyclooctane
SMILES C1CCCCCCC1

Specifications

Melting Point 12°C to 14°C
Density 0.835
Boiling Point 150°C to 152°C
Flash Point 28°C (82°F)
Refractive Index 1.458
Quantity 50 mL
UN Number UN3295
Beilstein 1900349
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 112.22
Percent Purity ≥99%
Chemical Name or Material Cyclooctane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252336 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog NumberL15000.AK,L15000.AD
Lot Number10247580 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL15000.AK,L15000.AD
Lot Number10243287 Certificate TypeCertificate of Analysis Date08/09/2022 Catalog NumberL15000.AK,L15000.AD
Lot Number10242170 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog NumberL15000.AK,L15000.AD
Lot Number10211125 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL15000.AK,L15000.AD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclooctane, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.