Learn More
Cyclopropanecarbonitrile, 98%
Description
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Specifications
Specifications
Density | 0.905 |
Boiling Point | 134°C to 135°C |
Flash Point | 32°C (89°F) |
Odor | Recognizable |
Refractive Index | 1.421 |
Quantity | 50 g |
UN Number | UN3275 |
Beilstein | 635790 |
Solubility Information | Miscible with water. |
Formula Weight | 67.09 |
Show More |
Safety and Handling


- Cyclopropanecarbonitrile
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Cyclopropanecarbonitrile
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.