missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl cyanide, 98%, Thermo Scientific Chemicals

Product Code. 11995601
500 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5500-21-0
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
InChI Key AUQDITHEDVOTCU-UHFFFAOYSA-N
Synonym cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
PubChem CID 79637
IUPAC Name cyclopropanecarbonitrile
SMILES C1CC1C#N

Specifications

Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 133.0°C to 135.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5CN
Refractive Index 1.4200 to 1.4220
Quantity 500 g
Beilstein 09,4
Specific Gravity 0.91
Formula Weight 67.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopropyl cyanide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopropyl cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopropyl cyanide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.