Learn More
(±)-3-Chloro-1,2-propane-1,1,2,3,3-d5-diol, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
CAS | 342611-01-2 |
Packaging | Glass Bottle |
Quantity | 25 mg |
Boiling Point | 160°C |
Flash Point | 113°C |
For Use With (Application) | Aliphatics |
Physical Form | Liquid |
Grade | Analytical Standard |