Learn More
Kartell Cylindrical Test Tube with Screw Cap, Polystyrene
Cylindrical test tubes equipped with a tight screw cap.
Brand:  Kartell 8820500
 View more versions of this product
Product Code. 18008041
  • 1629.78 DKK / Pack of 750
Explore more special offers
Description
Description
  • Test tubes are intended to contain blood and biological liquids to perform laboratory tests
Specifications
Specifications
Polypropylene
120 mm
Cylindrical
Non-sterile
No
Test Tube
15 mL
Polystyrene
Single-use
Not Autoclavable
Screw Cap