missing translation for 'onlineSavingsMsg'
Learn More
Cypermethrin, 95%, Thermo Scientific™ Product Code.: 15457179

Cypermethrin, 95%, Thermo Scientific™

Product Code. 15457179
1g
Click to view available options
Unit Size:
1g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 15457179

Brand: Thermo Scientific Alfa Aesar J65998.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Potent inhibitor of calcineurin; a pyrethrin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cypermethrin is used as pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin.

Solubility
Cypermethrin is soluble in methanol or acetone. The product is insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 52315-07-8
Flash Point 100°C (212°F)
Molecular Formula C22H19Cl2NO3
MDL Number MFCD00055328
UN Number UN2811
Merck Index 14,2767
Synonym alpha-Cyano-(3-phenoxyphenyl)methyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
InChI Key KAATUXNTWXVJKI-UHFFFAOYSA-N
SMILES CC1(C(C1C(=O)OC(C#N)C2=CC(=CC=C2)OC3=CC=CC=C3)C=C(Cl)Cl)C
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 416.298
PubChem CID 2912
ChEBI CHEBI:4042
Formula Weight 416.3
Percent Purity 95%
Chemical Name or Material Cypermethrin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberI20Z047 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberW30A048 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot NumberH03Y003 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot NumberR05A046 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot NumberZ22B051 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cypermethrin, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.