missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 18 Mouse anti-Human, Mouse, Rat, Clone: 1G11C4, Proteintech

Mouse Monoclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen Cytokeratin 18
Clone 1G11C4
Concentration 0.87 mg/mL
Applications Immunocytochemistry, Flow Cytometry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16847835
View Documents
Proteintech
66187-1-IG-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16837835
View Documents
Proteintech
66187-1-IG-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications

Specifications

Cytokeratin 18
0.87 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CK 18, CYK18, Cytokeratin 18, K18, keratin 18, KRT18
KRT18
IgG1
Protein G
Antibody
1G11C4
Immunocytochemistry, Flow Cytometry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Mouse, Rat
P05783, P05784, Q5BJY9
16668, 294853, 3875
Cytokeratin 18 Fusion Protein Ag1260
Primary
-20°C
KRT18
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytokeratin 18 Mouse anti-Human, Mouse, Rat, Clone: 1G11C4, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.