Learn More
Epredia™ Cytoslide™ Microscope Slides
Improve processing and visualization of small-volume specimens with these cytoslides, for use with Epredia™ Shandon™ EZ Single Cytofunnels™.
Brand: Epredia™ 5991051
1248.88 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Uncoated slides available with circle on front or back
- Available in cases of 100 slides
Specifications
Single Cytoslide | |
Plain | |
Shandon™ Single Cytoslides™; Noncoated, circle on back |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Consumables Monthly Promotion
Explore exclusive monthly deals on essential consumables for unbeatable savings.Promo Code: 24224Expires: 2025-05-31
New Offer Save Up to 74%