missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic beta-Glucosidase/GBA3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99838UV
This item is not returnable.
View return policy
Description
Cytosolic beta-Glucosidase/GBA3 Polyclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for ELISA.Specifications
Cytosolic beta-Glucosidase/GBA3 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 350 | |
CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
Produced in rabbits immunized with purified, recombinant Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met 1-Leu 469). | |
0.1 mL | |
metabolism | |
57733 | |
Store at 4°C in the dark. | |
IgG |