Learn More
D-Cystine 98%, Thermo Scientific Chemicals
174.78 DKK - 477.49 DKK
Chemical Identifiers
CAS | 349-46-2 |
---|---|
Molecular Formula | C6H12N2O4S2 |
Molecular Weight (g/mol) | 240.29 |
MDL Number | MFCD00002610 |
InChI Key | LEVWYRKDKASIDU-IMJSIDKUSA-N |
Synonym | d-cystine, 2-amino-3-2s-2-amino-2-carboxyethyl disulfanyl propanoic acid |
PubChem CID | 6857538 |
ChEBI | CHEBI:35494 |
IUPAC Name | (2S)-2-amino-3-[[(2S)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid |
SMILES | N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
349-46-2 | |
240.29 | |
LEVWYRKDKASIDU-IMJSIDKUSA-N | |
6857538 | |
(2S)-2-amino-3-[[(2S)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid |
C6H12N2O4S2 | |
MFCD00002610 | |
d-cystine, 2-amino-3-2s-2-amino-2-carboxyethyl disulfanyl propanoic acid | |
CHEBI:35494 | |
N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O |
Specifications
260.0°C | |
White | |
C6H12N2O4S2 | |
[-SCH2CH(NH2)CO2H]2 | |
15, 2779 | |
Solubility in water: 0.057g/L (25°C). | |
N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O | |
+ 214.00 | |
6857538 | |
240.29 | |
0.1% max. | |
D-Cystine, 98% |
349-46-2 | |
98% | |
Authentic | |
MFCD00002610 | |
d-cystine, 2-amino-3-2s-2-amino-2-carboxyethyl disulfanyl propanoic acid | |
LEVWYRKDKASIDU-IMJSIDKUSA-N | |
(2S)-2-amino-3-[[(2S)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid | |
240.29 | |
CHEBI:35494 | |
98% | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 206-486-2
RUO – Research Use Only