Learn More
Thermo Scientific Chemicals D-Threonine, 99% Product Code.: 11404677
Thermo Scientific Chemicals D-Threonine, 99%
Product Code. 11404677
100 g, 100g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11404677
Brand: Thermo Scientific Chemicals B21177.22
Request Bulk or Custom Format
CAS: 632-20-2 | C4H9NO3 | 119.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dthShow MoreShow Less
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O
Specifications
Melting Point ∼274°C (decomposition)
Beilstein 1721643
Quantity 100 g
Formula Weight 119.12
Optical Rotation +29° (c=5 in Water)
Percent Purity 99%
Chemical Name or Material D-Threonine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10251017 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeB21177.22,B21177.06,B21177.14
Lot Number10244097 Certificate TypeCertificate of Analysis Date20/10/2022 Product CodeB21177.22,B21177.06,B21177.14
Lot NumberT06I012 Certificate TypeCertificate of Analysis Date19/07/2022 Product CodeB21177.22,B21177.06,B21177.14
Lot Number10179593 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21177.22,B21177.06,B21177.14
Lot NumberH27Y052 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21177.22,B21177.06,B21177.14
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals D-Threonine, 99% >