missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDR1 Human Protein
DDR1 is a receptor tyrosine kinase activated by collagen.
13290.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_001945 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Molecular Weight (g/mol) | 78.1kDa |
Storage Requirements | Kinase proteins are shipped on dry ice and should be stored at -80°C. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468493
|
Invitrogen™
PR6913A |
100 μg |
13290.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12083755
|
Invitrogen™
PV6048 |
1 mg |
57500.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDR1 is reported to regulate adhesion, cell migration and other cellular processes.
Specifications
NP_001945 | |
78.1kDa | |
RUO - research use only | |
DDR1 | |
Human | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
DDR Kinase Family | |
GST Tag | |
Frozen |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title