Learn More
Mettler Toledo™ Dedicated Cable
Brand: Mettler Toledo™ 30027776
1290.75 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Mini-LTW | |
Dedicated Cable | |
InLab™ Smart Pro-ISM to Seven Compact S220 | |
1.2 m |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
Days
33Hours
19Minutes
3Seconds
42New Offer Save Now
Product Content Correction
Product Title
Mettler Toledo™ Dedicated Cable > Length Cable: 1.2m
Spot an opportunity for improvement?Share a Content Correction