missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEFA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18057-1-AP-150UL
This item is not returnable.
View return policy
Description
Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 3 by only one amino acid. This gene and the gene encoding defensin, alpha 3 are both subject to copy number variation. Two transcript variants encoding different isoforms have been found for this gene.Specifications
DEFA1 | |
Polyclonal | |
Unconjugated | |
DEFA1 | |
DEF1, DEFA1, DEFA1B, DEFA2, defensin, alpha 1, HNP 1, HNP 2, HP 1, HP 2, HP1, HP2, MRS, Neutrophil defensin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1667 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59665 | |
DEFA1 | |
DEFA1 Fusion Protein Ag12592 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DEFA1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction