Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (EU)
Bench Scales for Basic Industrial Applications
Brand: OHAUS™ 30685253
4849.62 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery | |
Standard RS232 and optional USB Device or Ethernet | |
15 kg | |
355 x 305 mm | |
D33XW15B1R5 | |
Weighing, Checkweighing, Counting, Totalization. | |
11.4 kg |
AC Adapter (Included);Rechargeable Battery (Included) | |
Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet | |
7-segment Multi-color Backlit LCD | |
2 g | |
-10°C to +40°C, 85%RH, non-condensing | |
646 x 305 x 122 mm |
Product Content Correction
Product Title
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (EU) > Bench Scale i-D33XW15B1R5-EU
Spot an opportunity for improvement?Share a Content Correction