Learn More
OHAUS™ Defender™ 5000 R Series Base
OHAUS™ Defender™ 5000 R Series Base offers durability for demanding industrial applications. The R series features moisture-protected single-point load cells, powder-coated steel construction, non-slip rubber adjustable feet, removable stainless steel weighing pans and leveling bubbles.
Brand: OHAUS™ 30419608
4812.32 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- The R Series features a stainless steel platform, powder-coated steel frame and an aluminum load cell to withstand harsh industrial environments.
- The Defender 5000 bases are equipped with an EasyConnect cable, which allows instant configuration with Defender 5000 (TD52) indicators, speeding up installation.
- Defender 5000 bases are NTEP-certified and Measurement Canada approved.
- These bases also carry OIML and EC Type approvals, which make them ideal for legal-for-trade use.
Specifications
304-grade stainless steel platform, sand grey powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet, level indicator | |
2.000000 g, 5.000000 g | |
IP67 | |
428 x 400 x 120 mm |
30.00 kg, 60.00 kg | |
D60RQDL | |
-10°C to +40°C | |
8.600 kg |