Learn More
Delgocitinib, MedChemExpress
MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Brand:  Medchem Express HY-109053
Encompass_Preferred
 View more versions of this product
Product Code. 16421129
  • 2083.52 DKK / 1mg
Explore more special offers
Description
Description
Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Specifications
Specifications
Delgocitinib
Off-White
C16H18N6O
JTE-052
O=C(N(C[C@@H]1C)[C@]21CN(C3=C(C=CN4)C4=NC=N3)CC2)CC#N
310.35
Research
COVID-19-immunoregulation
1263774-59-9
Research
1 mg
DMSO : ≥ 58 mg/mL (186.89 mM)
310.35
99.75%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.