Learn More
Desiccant Packs, Cytiva
Ensure that DNA sampling cards remain dry during transportation or storage
Brand: Whatman products Cytiva 10548239
207.77 DKK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
This item is not returnable.
View return policy
Description
- High vapor transmission rate
- Substrate made with Tyvek™ material
- Non-dusting, strong and hard to tear
Specifications
Tyvek™ Substrate | |
100 Pk./Cs. | |
1 g |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Product Title
Desiccant Packs, Cytiva > 1 g
Spot an opportunity for improvement?Share a Content Correction