missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl sulfide, 98+%

Product Code. 11476386
100 g, 100g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 111-47-7 | C6H14S | 118.24 g/mol

Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

Solubility
Soluble in water 351mg/L @ 25°C.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-47-7
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00009379
InChI Key ZERULLAPCVRMCO-UHFFFAOYSA-N
Synonym propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane
PubChem CID 8118
IUPAC Name 1-propylsulfanylpropane
SMILES CCCSCCC

Specifications

Melting Point -102°C
Density 0.838
Boiling Point 141°C to 142°C
Flash Point 28°C (82°F)
Odor Stench
Refractive Index 1.4487
Quantity 100 g
UN Number UN1993
Beilstein 1719002
Merck Index 14,7868
Solubility Information Soluble in water 351mg/L @ 25°C.
Formula Weight 118.24
Percent Purity ≥98%
Chemical Name or Material Di-n-propyl sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-n-propyl sulfide, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.