Learn More
MP Biomedicals Diagnostic Kit for Procalcitonin (PCT) (MPQuanti™ only)
Shop All MP Biomedical Products
Click to view available options
:
Procalcitonin
Description
- Diagnostic Kit for Procalcitonin (PCT) (Whole blood/serum/plasma) is based on fluorescence immunoassay technology.
- The sample moves through the strip from sample pad to absorbent pad. If the specimen contains Procalcitonin, it attaches to the fluorescent microspheres-conjugated anti-procalcitonin antibodies.
- Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
- The concentration of PCT in the sample correlates with the fluorescence signal intensity captured on the T line.
- According to the fluorescence intensity of the test and the standard curve, the concentration of PCT in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer to show PCT concentration in the sample.
- CE marked
- For professional in-vitro diagnostic use
- Working only with MPQuanti™ Immunofluorescence Analyzer
- Linear working range: 1-50 ng/mL, R≥0.990
- Test results should be interpreted at 15 minutes
- Used as an aid in the diagnosis of inflammatory conditions
Specifications
Specifications
Detectable Analytes | Procalcitonin |
Detection Method | Immunofluorescence |
Sample Type | Human whole blood, Human serum, Human plasma |
Type | POC, Quantitative, procalcitonin, PCT |
Form | Ready-to-Use (RTU) |
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for Procalcitonin (PCT) (MPQuanti™ only) >
Spot an opportunity for improvement?Share a Content Correction