Learn More
Diamond powder, natural, 40-60 micron
Description
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Chemical Name or Material | Diamond powder |
Name Note | Natural |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00211867 |
Percent Purity | 99.9% |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A24Y002 | Certificate of Analysis | 08/12/2021 | 038993.K2,038984.06,038984.03 |
K03X028 | Certificate of Analysis | 07/12/2021 | 038993.K2,038984.06,038984.03 |
J25P39 | Certificate of Analysis | 07/12/2021 | 038993.K2,038984.06,038984.03 |
J11Q038 | Certificate of Analysis | 29/11/2021 | 038993.K2,038984.06,038984.03 |
G31L02 | Certificate of Analysis | 24/11/2021 | 038993.K2,038984.06,038984.03 |