missing translation for 'onlineSavingsMsg'
Learn More
Diethylene Glycol, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10448020

Diethylene Glycol, Certified AR for Analysis, Fisher Chemical™

Product Code. 10448020
2.5 L
Click to view available options
Quantity:
500 mL
2.5 L
This item is not returnable. View return policy

Product Code. 10448020

Brand: Fisher Chemical D/2951/17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 111-46-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00002882
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digolShow MoreShow Less
PubChem CID 8117
ChEBI CHEBI:46807
IUPAC Name 2-(2-hydroxyethoxy)ethanol
SMILES OCCOCCO

Specifications

Melting Point -10°C
Color Colorless
Boiling Point 245°C
Packaging Amber glass bottle
Quantity 2.5 L
Formula Weight 106.12g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Diethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylene Glycol, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.