missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic container
Specifications
Specifications
Melting Point | -10°C |
Color | Colorless |
Boiling Point | 245°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
IUPAC Name | 2-(2-hydroxyethoxy)ethanol |
Formula Weight | 106.12g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Diethylene glycol
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction