missing translation for 'onlineSavingsMsg'
Learn More
Diethylgermanium dichloride, Thermo Scientific Chemicals Product Code.: 11307788

Diethylgermanium dichloride, Thermo Scientific Chemicals

Product Code. 11307788
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11307788

Brand: Thermo Scientific Alfa Aesar 034108.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As an intermediate

As an intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13314-52-8
Molecular Formula C4H14Cl2Ge
Molecular Weight (g/mol) 205.69
MDL Number MFCD00013589
InChI Key FVUWAWRMMUJVGO-UHFFFAOYSA-N
Synonym diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermaneShow MoreShow Less
PubChem CID 83334
IUPAC Name dichloro(diethyl)germane
SMILES Cl.Cl.CC[GeH2]CC

Specifications

Melting Point -37°C
Color Colorless
Density 1.372 g/mL
Boiling Point 175°C
Flash Point 63°C (145°F)
Odor Acrid
Refractive Index 1.471
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Decomposes in water and NH3; Soluble in organic solvents
Formula Weight 201.64
Physical Form Liquid
Chemical Name or Material Diethylgermanium dichloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20K035 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number034108.14,034108.06
Lot NumberS23J027 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number034108.03,034108.14,034108.06
Lot NumberY24H052 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number034108.14,034108.06
Lot NumberG30Y043 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number034108.14,034108.06
Lot NumberD20J28 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number034108.14,034108.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylgermanium dichloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.