Learn More
BD Difco™ Poly Antisera
For agglutination test
Brand:  BD 228371
 View more versions of this product
Product Code. 11772894
  • 2905.00 DKK / 3mL
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Description
Salmonella poly antiserum
  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For in vitro diagnostic use
Shigella
3 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Vial Shigella Antis Poly D Lyophilize
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide
For in vitro diagnostic use