Learn More
Karl Hecht™ Assistent™ Digital Interval Timers
Excellent for various timing purposes in the laboratory. Karl Hecht™ Assistent™ Digital Interval Timers are programmable up to 99 minutes and 59 seconds.
246.00 DKK
Description
- With magnetic back
- Includes battery
- Counts down in seconds
- Memory recall function
- Individual setting for minutes/seconds
- LCD display, 4 digits showing minutes/seconds
- Alarm sounds for 30 seconds when countdown reaches zero
Specifications
Digital |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Assistent™ Digital Interval Timers