Learn More
Dihydrotanshinone I, MedChemExpress
MedChemExpress Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.
Brand:  Medchem Express HY-N0360
Encompass_Preferred
 View more versions of this product
Product Code. 16483856
  • 2618.51 DKK / 25mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.
Specifications
Specifications
Dihydrotanshinone I
Red
C18H14O3
DMSO : 2 mg/mL (7.19 mM; Need ultrasonic)
278.3
98.74%
Powder
87205-99-0
Research
25 mg
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC[C@@H]4C)=O
278.3
Research
COVID-19-anti-virus
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.